Skip to Content
Merck
All Photos(1)

Key Documents

763535

Sigma-Aldrich

5-(3-Pyridyl)-1H-tetrazole

97%

Synonym(s):

3-(1H-Tetrazol-5-yl)pyridine, 5-β-Pyridyltetrazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5N5
CAS Number:
Molecular Weight:
147.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

238-242 (Decomposition)

SMILES string

c1cncc(c1)-c2nnn[nH]2

InChI

1S/C6H5N5/c1-2-5(4-7-3-1)6-8-10-11-9-6/h1-4H,(H,8,9,10,11)

InChI key

SECHDFHDDVELCV-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tian Wen et al.
Inorganic chemistry, 52(1), 12-14 (2012-12-19)
Two isomeric two-dimensional copper(I) coordination polymer materials based on an in situ generated 5-(3-pyridyl)tetrazole ligand show similar layer structures but distinct photoluminescent and photocatalytic properties, which present an interesting comparative study on the structure-property correlation between isomeric materials.
G D Stratton et al.
Biochemical pharmacology, 34(2), 275-279 (1985-01-15)
Using an isolated rat epididymal adipocyte system we have studied the development of tolerance to and cross-tolerance between nicotinic acid, 5-methylpyrazole-3-carboxylic acid and pyridyl-3-tetrazole. Preincubating isolated adipocytes with any one of these compounds results in a reduction of the antilipolytic
D D Myles et al.
Biochemical pharmacology, 34(2), 269-274 (1985-01-15)
The development of tolerance to the action of certain antilipolytic agents has been investigated in vivo in rats. Tolerance to oral nicotinic acid developed during twice daily dosing for 4 days at 100 and 250 mg/kg but not at 10

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service