Skip to Content
Merck
All Photos(1)

Documents

762660

Sigma-Aldrich

N-(2-Naphthyl)-1-naphthylamine

95%

Synonym(s):

N-(1-Naphthyl)-2-naphthylamine, N-(Naphthalen-2-yl)naphthalen-1-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H15N
CAS Number:
Molecular Weight:
269.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

form

solid

mp

102-107 °C

SMILES string

N(c1ccc2ccccc2c1)c3cccc4ccccc34

InChI

1S/C20H15N/c1-2-8-17-14-18(13-12-15(17)6-1)21-20-11-5-9-16-7-3-4-10-19(16)20/h1-14,21H

InChI key

UNJZLNFHHINVOB-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service