Skip to Content
Merck
All Photos(1)

Key Documents

762172

Sigma-Aldrich

Diethyl 3-bromobenzylphosphonate

97%

Synonym(s):

Diethyl m-bromobenzylphosphonate, [(3-Bromophenyl)methyl]phosphonic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16BrO3P
CAS Number:
Molecular Weight:
307.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.523

density

1.343 g/mL at 25 °C

functional group

bromo
phosphonate

SMILES string

CCOP(=O)(Cc1cccc(Br)c1)OCC

InChI

1S/C11H16BrO3P/c1-3-14-16(13,15-4-2)9-10-6-5-7-11(12)8-10/h5-8H,3-4,9H2,1-2H3

InChI key

KVNNRCDDRDXWIW-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service