Skip to Content
Merck
All Photos(1)

Documents

756458

Sigma-Aldrich

3-Buten-2-one-13C4

99 atom % 13C, 97% (CP), contains hydroquinone and acetic acid as stabilizer

Synonym(s):

Methyl vinyl ketone-13C4, Vinyl methyl ketone-13C4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C4H6O
Molecular Weight:
74.06
UNSPSC Code:
12352101

isotopic purity

99 atom % 13C

Assay

97% (CP)

form

liquid

contains

hydroquinone and acetic acid as stabilizer

bp

34 °C/120 mmHg (lit.)

density

0.722 g/mL at 25 °C

mass shift

M+4

storage temp.

2-8°C

SMILES string

[13CH3][13C]([13C]([H])=[13CH2])=O

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Flash Point(F)

19.4 °F

Flash Point(C)

-7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service