Skip to Content
Merck
All Photos(1)

Documents

754234

Sigma-Aldrich

Dichlorobis[(2,6-diisopropylphenyl)imido](1,2-dimethoxyethane)molybdenum(VI)

Synonym(s):

Bis(N-2,6-diisopropylaniline)dimethoxyethane molybdenum dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H44Cl2MoN2O2
CAS Number:
Molecular Weight:
607.51
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

solid

reaction suitability

core: molybdenum
reagent type: catalyst

mp

130-135 °C

SMILES string

COCCOC.CC(C)c1cccc(C(C)C)c1N=[Mo](Cl)(Cl)=Nc2c(cccc2C(C)C)C(C)C

InChI

1S/2C12H17N.C4H10O2.2ClH.Mo/c2*1-8(2)10-6-5-7-11(9(3)4)12(10)13;1-5-3-4-6-2;;;/h2*5-9H,1-4H3;3-4H2,1-2H3;2*1H;/q;;;;;+2/p-2

InChI key

HYZBNVVVUKPSEN-UHFFFAOYSA-L

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service