Skip to Content
Merck
All Photos(1)

Key Documents

753874

Sigma-Aldrich

CdSeS/ZnS alloyed quantum dots

COOH functionalized, fluorescence λem 575 nm, 6 nm diameter, 1 mg/mL in H2O

Synonym(s):

QD carboxyl functionalized, Fluorescent Nanocrystals, Trilite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdSxSe1-x/ZnS
UNSPSC Code:
26111700
NACRES:
NA.23
To inquire about this product 753874, please contact your local Merck office or dealer. Contact Technical Service

form

liquid

concentration

1 mg/mL in H2O

refractive index

n20/D 1.496 (lit.)

diameter

6 nm

bp

110-111 °C (lit.)

fluorescence

λem 575 nm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

These Photo and Thermostable Quantum Dot materials have comparable fluorescence and molecular kinetics across all colors. Narrow and sharp emission peaks result in less overlap of peaks when multiplexing.

Analysis Note

Full Width at Half Maximum: 30 - 40nm

Legal Information

For use in optical electronics research only.
Product of CytoDiagnostics
Trilite is a trademark of Cytodiagnostics Inc.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Dye-sensitized solar cells as a promising, low-cost photovoltaic technology.

Professor Sharma and colleagues review the synthesis and applications of this novel material. This includes a discussion of the unique properties of quantum dots and their suitability for solar cell applications, along with common synthesis techniques used to develop these materials.

Professor Xiaohu Gao (University of Washington, USA) provides a overview of recent quantum dot (QD) advancements and their potential for advancing bioassay and bioimaging technologies.

Perovskite quantum dots research progresses overcoming challenges, enabling rapid development of light-emitting devices.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service