Skip to Content
Merck
All Photos(2)

Key Documents

748188

Sigma-Aldrich

Bis(2,2,6,6-tetramethylpiperidinyl)zinc, lithium chloride, magnesium chloride complex solution

in THF/toluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C18H36N2Zn · xMgCl2 · xLiCl
UNSPSC Code:
12352103
NACRES:
NA.22

form

liquid

composition

Bis(2,2,6,6-tetramethylpiperidinyl)zinc, ~12 wt. %
LiCl, 2-4%
MgCl2, 6-9%
THF/toluene, ca. 90:10 (solvent)

concentration

in THF/toluene

density

0.991 g/mL at 25 °C

General description

Bis(2,2,6,6-tetramethylpiperidinyl)zinc,lithium chloride, magnesium chloride complex solution is an organozinc reagentextensively used in C-C bond forming,Negishi coupling, Michael additions, Cu(I)-promoted reactions, and electrophilic amination reactions.

Application

Bis(2,2,6,6-tetramethylpiperidinyl)zinc, lithium chloride, magnesium chloride complex solution can be used as a base for the:
  • Regioselective zincation of pyridines and related azines to synthesize zincated pyridines and azines.(1)
  • Chemoselective direct zincation of arenes and heteroarenes to synthesize polyfunctional aryl and heteroaryl zinc reagents.
  • Regioselective zincation of thiazoles to synthesize zincated thiazoles, which readily reacts with various electrophiles to form highly functionalized thiazoles.

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

5.0 °F

Flash Point(C)

-15 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service