Skip to Content
Merck
All Photos(2)

Documents

740942

Sigma-Aldrich

Indium antimonide

≥99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
InSb
CAS Number:
Molecular Weight:
236.58
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

≥99.99% trace metals basis

form

crystals

SMILES string

[In]#[Sb]

InChI

1S/In.Sb

InChI key

WPYVAWXEWQSOGY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Indium antimonide (InSb) is a semiconductor material with good electron mobility and saturation velocity. It can be prepared typically by ball-milling indium (In) and antimony (Sb) metals in a 1:1 ratio.

Application

InSb can be used for a variety of applications such as superconductors, thermoelectric materials, and lithium-ion batteries.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Signatures of Majorana fermions in hybrid superconductor-semiconductor nanowire devices
Mourik V, et al.
Science, 3(1), 13-13 (2012)
High performance In x Ce y Co 4 Sb 12 thermoelectric materials with in situ forming nanostructured InSb phase
Li H, et al.
Applied Physics Letters, 336(6084), 1003-1007 (2009)
Intermetallic Insertion Electrodes with a Zinc Blende-Type Structure for Li Batteries: A Study of Li x InSb
Vaughey JT, et al.
Electrochemical and Solid-State Letters, 121-124 (1999)
Electrochemistry and in-situ X-ray diffraction of InSb in lithium batteries
Johnson CS, et al.
Electrochemical Communications, 94(10), 102114-102114 (2000)
Fritzsche, H.; Lark-Horovitz, K.
Physical Review. B, Condensed Matter and Materials Physics, 99, 400-400 (1955)

Articles

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service