Skip to Content
Merck
All Photos(1)

Documents

740608

Sigma-Aldrich

tert-Butyl (tert-butyldimethylsilyl)glyoxylate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H24O3Si
CAS Number:
Molecular Weight:
244.40
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.440

density

0.920 g/mL at 25 °C

SMILES string

CC(C)(C)OC(=O)C(=O)[Si](C)(C)C(C)(C)C

InChI

1S/C12H24O3Si/c1-11(2,3)15-9(13)10(14)16(7,8)12(4,5)6/h1-8H3

InChI key

GGYHTNUVBJRLDM-UHFFFAOYSA-N

Application

Learn More at the Professor and Product Portal of Professor Jeffrey S. Johnson.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

190.9 °F

Flash Point(C)

88.3 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Johnson group has developed a family of reagents that are useful for the modular preparation of highly substituted glycolic acid derivatives. Silyl glyoxylates are potent electrophiles by virtue of being α-dicarbonyls, and react with a variety of nucleophiles.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service