Skip to Content
Merck
All Photos(1)

Key Documents

73476

Sigma-Aldrich

(SP)-1-[(S)-α-(Dimethylamino)-2-(diphenylphosphino)benzyl]-2-diphenylphosphinoferrocene

≥97%, optical purity ee: ≥99%

Synonym(s):

(2R)-1-[(S)-(Dimethylamino)[2-(diphenylphosphino)phenyl]methyl]-2-(diphenylphosphino)ferrocene (acc to CAS), Taniaphos SL-T001-2

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CZK 1,659.00
500 MG
CZK 11,500.00
1 G
CZK 18,600.00
5 G
CZK 86,000.00

CZK 1,659.00

List priceCZK 2,370.00Save 30%

Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
CZK 1,659.00
500 MG
CZK 11,500.00
1 G
CZK 18,600.00
5 G
CZK 86,000.00

About This Item

Empirical Formula (Hill Notation):
C43H39FeNP2
CAS Number:
Molecular Weight:
687.57
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

CZK 1,659.00

List priceCZK 2,370.00Save 30%

Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥97%

form

powder

optical purity

ee: ≥99%

functional group

amine
phosphine

storage temp.

2-8°C

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.CN(C)[C@H]([C]2[CH][CH][CH][C]2P(c3ccccc3)c4ccccc4)c5ccccc5P(c6ccccc6)c7ccccc7

InChI

1S/C38H34NP2.C5H5.Fe/c1-39(2)38(35-27-17-29-37(35)41(32-22-11-5-12-23-32)33-24-13-6-14-25-33)34-26-15-16-28-36(34)40(30-18-7-3-8-19-30)31-20-9-4-10-21-31;1-2-4-5-3-1;/h3-29,38H,1-2H3;1-5H;/t38-;;/m0../s1

InChI key

DEHRMLQYTSYVBC-ZCLNGFLVSA-N

General description

sold in collaboration with Solvias AG

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service