Skip to Content
Merck
All Photos(1)

Key Documents

732001

Sigma-Aldrich

N-Methyl-13C,d3-ethanolamine

99 atom % 13C, 98 atom % D, 97% (CP)

Synonym(s):

2-(Methyl-13C, d3-amino)ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CD3NHCH2CH2OH
Molecular Weight:
79.12
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C
98 atom % D

Assay

97% (CP)

form

liquid

bp

159 °C (lit.)

density

0.984 g/mL at 25 °C

mass shift

M+4

SMILES string

[2H][13C]([2H])([2H])NCCO

InChI

1S/C3H9NO/c1-4-2-3-5/h4-5H,2-3H2,1H3/i1+1D3

InChI key

OPKOKAMJFNKNAS-KQORAOOSSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2 - Skin Corr. 1B - STOT RE 2 Oral - STOT SE 3

Target Organs

Kidney,Testes,Liver,spleen,ovary, Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

168.8 °F

Flash Point(C)

76 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service