Skip to Content
Merck
All Photos(2)

Key Documents

731439

Sigma-Aldrich

9,10-Bis[(triisopropylsilyl)ethynyl]anthracene

>99%

Synonym(s):

9,10-Bis{2-[tris(1-methylethyl)silyl]ethynyl}anthracene, TIPS-anthracene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H50Si2
CAS Number:
Molecular Weight:
538.95
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

>99%

form

solid

mp

211-214 °C

λmax

1.8732 at 274 nm

absorption

0.25821 at 394 nm
0.61112 at 416 nm
0.84016 at 442 nm

fluorescence

λex 435 nm; λem 475 nm (concentration = 0.00002 mg/mL)

Orbital energy

HOMO 5.6 eV (CV)
LUMO 2.9 eV (CV)

SMILES string

CC(C)[Si](C#Cc1c2ccccc2c(C#C[Si](C(C)C)(C(C)C)C(C)C)c3ccccc13)(C(C)C)C(C)C

InChI

1S/C36H50Si2/c1-25(2)37(26(3)4,27(5)6)23-21-35-31-17-13-15-19-33(31)36(34-20-16-14-18-32(34)35)22-24-38(28(7)8,29(9)10)30(11)12/h13-20,25-30H,1-12H3

InChI key

FEAZNDXDTUMTIL-UHFFFAOYSA-N

General description

9,10-Bis[(triisopropylsilyl)ethynyl]anthracene is a novel organic photocatalyst.

Application

The photocatalyst may be useful in the affording free radicals during polymerization via irradiated photoredox catalysis.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Organic photocatalyst for polymerization reactions: 9, 10-bis [(triisopropylsilyl) ethynyl] anthracene.
Tehfe, Mohamad-Ali, et al.
ACS Macro Letters, 198-203 null
Intermolecular Effects on the Hole States of Triisopropylsilylethynyl-Substituted Oligoacenes
Griffith, O. L.; Jones, A. G.; Anthony, J. E.; Lichtenberger, D. L.
The Journal of Physical Chemistry C, 114, 13838-13838 (2010)
Organic photocatalyst for polymerization reactions: 9, 10-bis [(triisopropylsilyl) ethynyl] anthracene.
Tehfe, Mohamad-Ali, et al.
ACS Macro Letters null
Jong-Hwa Park et al.
Organic letters, 9(13), 2573-2576 (2007-06-05)
New soluble anthracene derivatives containing thiophene and phenylenevinylene derivatives were synthesized via well-known synthetic routes. TIPS derivatives were added at the 9,10-positions of anthracene for the solubility and crystallinity. Both of the molecules were found to be promising for high

Articles

Organic materials in optoelectronic devices like LEDs and solar cells are of significant academic and commercial interest.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service