Skip to Content
Merck

Skip To

73028

N-Succinimidyl 4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-heptadecafluoroundecanoate

≥95%

Synonym(s):

N-Succinimidyl 2H,2H,3H,3H-perfluoroundecanoate, N-Succinimidyl 3-(perfluorooctyl)propionate

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C15H8F17NO4
CAS Number:
Molecular Weight:
589.20
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
eCl@ss:
32160702
Assay:
≥95%
Form:
crystals


assay

≥95%

form

crystals

mp

140-146 °C

storage temp.

−20°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCC(=O)ON1C(=O)CCC1=O

InChI

1S/C15H8F17NO4/c16-8(17,4-3-7(36)37-33-5(34)1-2-6(33)35)9(18,19)10(20,21)11(22,23)12(24,25)13(26,27)14(28,29)15(30,31)32/h1-4H2

InChI key

PFSIGNUPTCQGSG-UHFFFAOYSA-N

Legal Information

Product of Fluorous Technologies, Inc. Use of this compound may be protected by U.S. patents 6,156,896; 5,859,247; 5,777,121 and 6,673,539.


Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1

Target Organs

Liver

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters