Skip to Content
Merck
All Photos(1)

Key Documents

726648

Sigma-Aldrich

(R)-(+)-1-(1-Naphthyl)ethylamine

ChiPros®, produced by BASF, 99%

Synonym(s):

(R)-(+)-α-Methyl-1-naphthalenemethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7CH(CH3)NH2
CAS Number:
Molecular Weight:
171.24
Beilstein:
2208025
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

produced by BASF

Assay

≥98.5% (GC)
99%

form

liquid

optical purity

enantiomeric excess: ≥98.5%

refractive index

n20/D 1.623 (lit.)

bp

153 °C/11 mmHg (lit.)

density

1.067 g/mL at 20 °C (lit.)

SMILES string

C[C@@H](N)c1cccc2ccccc12

InChI

1S/C12H13N/c1-9(13)11-8-4-6-10-5-2-3-7-12(10)11/h2-9H,13H2,1H3/t9-/m1/s1

InChI key

RTCUCQWIICFPOD-SECBINFHSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiPros is a registered trademark of BASF SE

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Chiral amines play an important role in stereoselective organic synthesis. They are used directly as resolving agents, building blocks, or chiral auxiliaries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service