Skip to Content
Merck
All Photos(1)

Key Documents

726583

Sigma-Aldrich

(S)-(−)-α-Methylbenzylamine

ChiPros®, produced by BASF, ≥99.0%

Synonym(s):

(S)-(-)-alpha-Methylbenzylamine, (S)-(−)-1-Phenylethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)NH2
CAS Number:
Molecular Weight:
121.18
Beilstein:
2204907
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22
To inquire about this product 726583, please contact your local Merck office or dealer. Contact Technical Service

grade

produced by BASF

vapor pressure

0.5 mmHg ( 20 °C)

Assay

≥99.0% (GC)
≥99.0%

form

liquid

optical purity

enantiomeric excess: ≥99.0%

refractive index

n20/D 1.526 (lit.)

bp

187 °C (lit.)

density

0.94 g/mL at 25 °C (lit.)

functional group

amine
phenyl

SMILES string

C[C@H](N)c1ccccc1

InChI

1S/C8H11N/c1-7(9)8-5-3-2-4-6-8/h2-7H,9H2,1H3/t7-/m0/s1

InChI key

RQEUFEKYXDPUSK-ZETCQYMHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • The first example of a diastereoselective thio-Ugi reaction: a new synthetic approach to chiral imidazole derivatives.: This research presents a novel diastereoselective thio-Ugi reaction, enabling the synthesis of chiral imidazole derivatives, which are essential in asymmetric synthesis and pharmaceutical intermediates (Gulevich et al., 2007).
  • Synthesis and serotonin receptor affinities of a series of enantiomers of alpha-methyltryptamines: evidence for the binding conformation of tryptamines at serotonin 5-HT1B receptors.: This study investigates the synthesis and serotonin receptor affinities of alpha-methyltryptamine enantiomers, contributing to the understanding of chiral compounds in medicinal chemistry (Nichols et al., 1988).

Legal Information

ChiPros is a registered trademark of BASF SE

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bartosz Lewandowski et al.
Chemical communications (Cambridge, England), (47)(47), 6399-6401 (2008-12-03)
Simple chiral aza-crown ethers based on sucrose display high enantioselectivity in complexation of phenylethylammonium chlorides.
Andrew M Kelly et al.
Nature protocols, 3(2), 215-219 (2008-02-16)
A three-component chiral derivatization protocol for determining the enantiopurity of chiral diols by (1)H NMR spectroscopic analysis is described here. The present approach involves the derivatization of 1,2- 1,3- and 1,4-diols with 2-formylphenylboronic acid and enantiopure alpha-methylbenzylamine. This method affords
G Palaprat et al.
The journal of physical chemistry. B, 111(31), 9239-9243 (2007-07-20)
A polydomain cholesteric elastomer was obtained by cross-linking a nematic side-chain polysiloxane in the presence of a chiral dopant. After extraction of the chiral dopant, sorption experiments were performed, by the use of an electronic microbalance, in the presence of
Alejandra León et al.
Journal of natural products, 75(5), 859-864 (2012-05-12)
The enantiomeric lactams (-)-8, (+)-8, (+)-9, and (-)-9 were formed by the reaction of the dimeric phthalide rac-tokinolide B (rac-3) with (R)-(+)-α-methylbenzylamine and (S)-(-)-α-methylbenzylamine. The absolute configurations of compounds 8 and 9 were assigned by experimental and theoretically calculated electronic
Barry Lygo et al.
Organic & biomolecular chemistry, 10(25), 4968-4976 (2012-05-23)
A short synthetic route to diastereoisomeric atropos dihydro-5H-dibenz[c,e]azepinium salts via reaction of a single enantiomer of (R)-α-methylbenzylamine with a racemic atropos biphenol derivative is described. Compounds prepared via this approach are used to provide strong evidence that structurally related tropos

Articles

Chiral amines play an important role in stereoselective organic synthesis. They are used directly as resolving agents, building blocks, or chiral auxiliaries.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service