Skip to Content
Merck
All Photos(1)

Documents

726435

Sigma-Aldrich

(1R)-2,2′-Bis[(S)-(4-methylphenyl)sulfinyl]-1,1′-binaphthalene

97%

Synonym(s):

(M,S,S)-1,1′-Binaphthalene-2,2′-diyl-bis(p-tolylsulfoxide), (M,S,S)-pTol-BINASO, (R)-2,2′-Bis-((S)-toluene-4-sulfinyl)-[1,1′]binaphthalenyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H26O2S2
CAS Number:
Molecular Weight:
530.70
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.5% (HPLC)
97%

form

powder

optical purity

ee: ≥97.0%

SMILES string

Cc1ccc(cc1)S(=O)c2ccc3ccccc3c2-c4c(ccc5ccccc45)S(=O)c6ccc(C)cc6

InChI

1S/C34H26O2S2/c1-23-11-17-27(18-12-23)37(35)31-21-15-25-7-3-5-9-29(25)33(31)34-30-10-6-4-8-26(30)16-22-32(34)38(36)28-19-13-24(2)14-20-28/h3-22H,1-2H3/t37-,38-/m0/s1

InChI key

GAWRHNNPYUEUOO-UWXQCODUSA-N

Application

(1R)-2,2′-Bis[(S)-(4-methylphenyl)sulfinyl]-1,1′-binaphthalene (p-tolyl-BINASO) is a bis-sulfoxide ligand that can be used to prepare cationic palladium and platinum complexes. These complexes are employed as catalysts in hydroboration and diboration reactions. It can also be used to prepare the precatalyst [p-tol-BINASO}RhCl]2 for the synthesis of chiral arylcycloketones by the 1,4-addition reaction of arylboronic acids to cyclic α,β-unsaturated ketones.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, structure, and catalytic studies of palladium and platinum bis-sulfoxide complexes
Drinkel EE, et al.
Organometallics, 33(3), 627-636 (2014)
Disulfoxide ligands in rhodium catalyzed asymmetric 1,4-addition: first studies and future directions.
Mariz, R., et al.
Chimia, 63, 508-511 (2009)
A chiral bis-sulfoxide ligand in late-transition metal catalysis; rhodium-catalyzed asymmetric addition of arylboronic acids to electron-deficient olefins.
Ronaldo Mariz et al.
Journal of the American Chemical Society, 130(7), 2172-2173 (2008-01-29)
A chiral bis-sulfoxide ligand in late-transition metal catalysis; rhodium-catalyzed asymmetric addition of arylboronic acids to electron-deficient olefins
Mariz R, et al.
Journal of the American Chemical Society, 130(7), 2172-2173 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service