Skip to Content
Merck
All Photos(2)

Key Documents

716502

Sigma-Aldrich

[1,1′:4′,1″]Terphenyl- 3,3″,5,5″-tetracarboxylic acid

greener alternative

contains up to 2 equivalents of DMF

Synonym(s):

H4TPTC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H14O8
CAS Number:
Molecular Weight:
406.34
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

form

solid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

>370 °C

greener alternative category

SMILES string

OC(=O)c1cc(cc(c1)-c2ccc(cc2)-c3cc(cc(c3)C(O)=O)C(O)=O)C(O)=O

InChI

1S/C22H14O8/c23-19(24)15-5-13(6-16(9-15)20(25)26)11-1-2-12(4-3-11)14-7-17(21(27)28)10-18(8-14)22(29)30/h1-10H,(H,23,24)(H,25,26)(H,27,28)(H,29,30)

InChI key

HQCSLVYEWMDWIZ-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

[1,1′:4′,1″]Terphenyl- 3,3″,5,5″-tetracarboxylic acid (H4TPTC) is used as a linker for the synthesis of NOTT-101 metal organic framework (MOFs).

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selected Applications of Metal-Organic Frameworks in Sustainable Energy Technologies?.
Caskey SR and Matzger AJ.
Material Matters, 4(4), 111-111 (2009)

Articles

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service