Skip to Content
Merck
All Photos(1)

Documents

71334

Sigma-Aldrich

Sodium 1-butanethiolate

technical, ≥95.0% (RT)

Synonym(s):

1-Butanethiol sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH2SNa
CAS Number:
Molecular Weight:
112.17
Beilstein:
3594199
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Quality Level

Assay

≥95.0% (RT)

form

powder

SMILES string

CCCCS[Na]

InChI

1S/C4H10S.Na/c1-2-3-4-5;/h5H,2-4H2,1H3;/q;+1/p-1

InChI key

VTLWRSNWWLIJTQ-UHFFFAOYSA-M

Other Notes

Salts of alkanethiols are versatile intermediates for the synthesis of various thiol esters and thioethers. In contrast to the stenchy gaseous or liquid thiols, the salts are easy to handle solids.; Alkanethiolates are strong nucleophiles and thus efficient reagents for SN2 type dealkylations of hindered esters and lactones; Cleavage of aryl methyl ethers

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L. Testaferri et al.
Synthesis, 751-751 (1983)
G.I. Feutrill et al.
Tetrahedron Letters, 1327-1327 (1970)
P.A. Bartlett et al.
Tetrahedron Letters, 4459-4459 (1970)
W.R. Vaughan et al.
The Journal of Organic Chemistry, 27, 739-739 (1962)
Jung Bae Lee et al.
Ultramicroscopy, 108(10), 1352-1355 (2008-06-21)
Oligophenylethynylene thiol containing carboxylic acid in the tail group as a conducting wire bioreceptor was synthesized, and then its electrical property was investigated from the measurement of scanning tunneling microscopy (STM). Mixed self-assembled monolayer (SAM) consisting of 4-(2-(4-acetylthio)phenyl)ethynyl) benzoic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service