Skip to Content
Merck
All Photos(1)

Documents

710636

SAFC

(11bR)-4,5-Dihydro-3H-dinaphtho[2,1-c:1′,2′-e]phosphepine borane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20BP
Molecular Weight:
326.18
UNSPSC Code:
12352209
PubChem Substance ID:

Assay

97%

form

solid

optical activity

[α]22/D −272°, c = 0.5 in chloroform

mp

201-205 °C

SMILES string

B.C1PCc2ccc3ccccc3c2-c4c1ccc5ccccc45

InChI

1S/C22H17P.BH3/c1-3-7-19-15(5-1)9-11-17-13-23-14-18-12-10-16-6-2-4-8-20(16)22(18)21(17)19;/h1-12,23H,13-14H2;1H3

InChI key

CDJLGSCFLBPQSK-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Sol. 2 - Resp. Sens. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service