Skip to Content
Merck
All Photos(1)

Key Documents

710466

Sigma-Aldrich

Bis(2-methoxyphenyl)phosphine

97%

Synonym(s):

Di-o-anisylphosphine

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
CZK 2,560.00

CZK 2,560.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
CZK 2,560.00

About This Item

Empirical Formula (Hill Notation):
C14H15O2P
CAS Number:
Molecular Weight:
246.24
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

CZK 2,560.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

powder

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

86-90 °C

functional group

phosphine

SMILES string

COc1ccccc1Pc2ccccc2OC

InChI

1S/C14H15O2P/c1-15-11-7-3-5-9-13(11)17-14-10-6-4-8-12(14)16-2/h3-10,17H,1-2H3

InChI key

HFEAMIKDDWKNAG-UHFFFAOYSA-N

Application

Reactant for:
  • Allylation of phenols
  • Synthesis of neutral rhodium(I) complexes
  • Dinuclear Pd(II) complexes bearing bis-bidentate tetraphos ligands
  • CO-ethylene copolymerization reactions
  • Suzuki-Miyaura reaction for synthesis of alkyl derivatives phosphine sulfonate (P-O) ligands

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service