Skip to Content
Merck
All Photos(1)

Key Documents

70590

Sigma-Aldrich

4-Hydroxy-1-naphthalenesulfonic acid sodium salt

technical, ~70% (T)

Synonym(s):

1-Naphthol-4-sulfonic acid sodium salt, Sodium 4-hydroxy-1-naphthalenesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7NaO4S
CAS Number:
Molecular Weight:
246.21
Beilstein:
3640154
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

~70% (T)

form

powder

SMILES string

[Na+].Oc1ccc(c2ccccc12)S([O-])(=O)=O

InChI

1S/C10H8O4S.Na/c11-9-5-6-10(15(12,13)14)8-4-2-1-3-7(8)9;/h1-6,11H,(H,12,13,14);/q;+1/p-1

InChI key

IAAKNVCARVEIFS-UHFFFAOYSA-M

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Bessette
Journal of biochemical and biophysical methods, 1(4), 209-219 (1979-08-01)
The differential mobilities of compounds in an electric field are important analytical criteria and we can use them to bring electrophoretically pure components of a mixture medium on which they are separated. To this end, the compound undergoing reaction are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service