Skip to Content
Merck

69874

Molybdenum(VI) fluoride

≥99.9%

Synonym(s):

Hexafluoromolybdenum, Molybdenum hexafluoride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
MoF6
CAS Number:
Molecular Weight:
209.93
UNSPSC Code:
12352302
PubChem Substance ID:
EC Number:
232-026-5
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥99.9%

bp

37 °C (lit.)

mp

17.5 °C (lit.)

density

2.3 g/mL at 25 °C (lit.)

SMILES string

F[Mo](F)(F)(F)(F)F

InChI

1S/6FH.Mo/h6*1H;/q;;;;;;+6/p-6

InChI key

RLCOZMCCEKDUPY-UHFFFAOYSA-H



pictograms

Skull and crossbonesCorrosion

signalword

Danger

hcodes

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Dam. 1 - Skin Corr. 1A

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library