Skip to Content
Merck
All Photos(1)

Documents

698199

Sigma-Aldrich

cis-1-Propenylboronic acid MIDA ester

Synonym(s):

cis-1-Propene-1-boronic acid MIDA ester, cis-6-Methyl-2-(1-propenyl)-1,3,6,2-dioxazaborocane-4,8-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12BNO4
CAS Number:
Molecular Weight:
197.00
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

powder

mp

99-110 °C

SMILES string

C\C=C/B1OC(=O)CN(C)CC(=O)O1

InChI

1S/C8H12BNO4/c1-3-4-9-13-7(11)5-10(2)6-8(12)14-9/h3-4H,5-6H2,1-2H3/b4-3-

InChI key

PKQGENZEEBZVIL-ARJAWSKDSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service