Skip to Content
Merck
All Photos(1)

Key Documents

698180

Sigma-Aldrich

Tris(trimethylsilyl)silyl vinyl ether

Synonym(s):

Tris(trimethylsilyl)silyloxyethylene, Vinyloxytris(trimethylsilyl)silane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H30OSi4
CAS Number:
Molecular Weight:
290.70
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

mp

51-61 °C

storage temp.

2-8°C

SMILES string

C[Si](C)(C)[Si](OC=C)([Si](C)(C)C)[Si](C)(C)C

InChI

1S/C11H30OSi4/c1-11-12-16(13(2,3)4,14(5,6)7)15(8,9)10/h11H,1H2,2-10H3

InChI key

WPZQENSANSNYSI-UHFFFAOYSA-N

Application

Super Silyl Protecting Groups

Used for:
  • Diastereoselective [2+2] cyclizations
  • Cross-Aldol cascade reaction
  • Diastereoselective sequential reactions for generating polyols
  • Accessing distinct diastereomers and 4-component reactions
  • Supersilyl-directed aldol reactions

  • Strong electron-donating effect of the supersilyl group operates only in the α-position and not in the β-position

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

172.0 °F

Flash Point(C)

77.8 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service