Skip to Content
Merck
All Photos(1)

Key Documents

697648

Sigma-Aldrich

(3-Hydroxy-1-propenyl)sulfur pentafluoride

technical grade

Synonym(s):

Pentafluoro(3-hydroxy-1-propenyl)sulfur

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5F5OS
CAS Number:
Molecular Weight:
184.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical grade

refractive index

n20/D 1.381

density

1.692 g/mL at 25 °C

SMILES string

OC\C=C\S(F)(F)(F)(F)F

InChI

1S/C3H5F5OS/c4-10(5,6,7,8)3-1-2-9/h1,3,9H,2H2/b3-1+

InChI key

MGHHQQCFLJIKOP-HNQUOIGGSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

185.0 °F

Flash Point(C)

85 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service