Skip to Content
Merck
All Photos(1)

Documents

696153

Sigma-Aldrich

p-Xylene-13C8

98% (CP), 99 atom % 13C

Synonym(s):

1,4-Dimethylbenzene-13C8

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13C6H4(13CH3)2
CAS Number:
Molecular Weight:
114.11
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

98% (CP)

form

liquid

refractive index

n20/D 1.495

bp

138 °C

mp

12-13 °C

density

0.925 g/mL at 20 °C

mass shift

M+8

SMILES string

[13CH3][13c]1[13cH][13cH][13c]([13CH3])[13cH][13cH]1

InChI

1S/C8H10/c1-7-3-5-8(2)6-4-7/h3-6H,1-2H3/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1

InChI key

URLKBWYHVLBVBO-LLXNCONNSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service