Skip to Content
Merck
All Photos(3)

Key Documents

687855

Sigma-Aldrich

N-Isopropylidene-N′-2-nitrobenzenesulfonyl hydrazine

97%

Synonym(s):

2-(1-Methylethylidene)hydrazide-2-nitrobenzenesulfonic acid, Acetone 2-nitrobenzenesulfonylhydrazone, IPNBSH, Isopropylidenehydrazide o-nitro-benzenesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11N3O4S
CAS Number:
Molecular Weight:
257.27
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Assay

97%

form

solid

reaction suitability

reagent type: reductant

mp

131-135 °C

functional group

amine
hydrazide
nitro

SMILES string

C\C(C)=N\NS(=O)(=O)c1ccccc1[N+]([O-])=O

InChI

1S/C9H11N3O4S/c1-7(2)10-11-17(15,16)9-6-4-3-5-8(9)12(13)14/h3-6,11H,1-2H3

InChI key

SBNYNTYNEJTMQO-UHFFFAOYSA-N

Application

IPNBSH A Reagent for the Simple Reduction of Alcohols

Reactant for synthesis of:
  • (-)-Acylfulvene and (-)-irofulven for use as antitumor agents
  • Bicycloundecadienones and bicyclodecadienones via carbonylative cycloaddition
  • Monoalkyldiazenes for allylic reduction

Reacts with di-Me phosphonate
Reagent for allylic transposition and reduction of alcohols, as well as mild and stereoselective reduction of allylic carbonates and vinyl epoxides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad Movassaghi et al.
The Journal of organic chemistry, 72(5), 1838-1841 (2007-02-06)
The reagent N-isopropylidene-N'-2-nitrobenzenesulfonyl hydrazine (IPNBSH) is used in the reduction of alcohols via the loss of dinitrogen from transiently formed monoalkyl diazene intermediates accessed by sequential Mitsunobu displacement, hydrolysis, and fragmentation under mild reaction conditions.
A stereospecific palladium-catalyzed route to monoalkyl diazenes for mild allylic reduction.
Mohammad Movassaghi et al.
Angewandte Chemie (International ed. in English), 47(46), 8909-8912 (2008-10-17)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service