Skip to Content
Merck
All Photos(1)

Documents

682292

Sigma-Aldrich

(S)-(+)-4,12-Bis[di(3,5-xylyl)phosphino]-[2.2]-paracyclophane

90%

Synonym(s):

(S)-5,11-Bis(3,5-xylylphosphino)tricyclo[8.2.24,7]hexadeca-hexaene, (S)-xylyl-PHANEPHos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H50P2
CAS Number:
Molecular Weight:
688.86
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

form

solid

optical activity

[α]/D +61.0°, c = 0.1 in ethanol

mp

234-238 °C

InChI

1S/C48H50P2/c1-31-17-32(2)22-43(21-31)49(44-23-33(3)18-34(4)24-44)47-29-39-9-13-41(47)15-11-40-10-14-42(16-12-39)48(30-40)50(45-25-35(5)19-36(6)26-45)46-27-37(7)20-38(8)28-46/h9-10,13-14,17-30H,11-12,15-16H2,1-8H3

InChI key

LYHOBZAADXPPNW-UHFFFAOYSA-N

Application

Efficient ligand for asymmetric hydrogenation of dehydroamino acids, methyl esters and ketones.

Legal Information

Sold in collaboration with Johnson Matthey Catalyst for research purposes only. US5874629 and any patents arising therefrom apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burk et al.
Organic letters, 2(26), 4173-4176 (2001-01-11)
PhanePhos-ruthenium-diamine complexes catalyze the asymmetric hydrogenation of a wide range of aromatic, heteroaromatic, and alpha, beta-unsaturated ketones with high activity and excellent enantioselectivity.
Journal of the American Chemical Society, 119, 6207-6207 (1997)

Articles

P-Phos ligand family enhances catalysis, featuring atropisomeric biaryl bisphosphine with unique structural elements.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service