Skip to Content
Merck
All Photos(1)

Key Documents

678678

Sigma-Aldrich

3-Noradamantanecarbonyl chloride

97%

Synonym(s):

3-Noradamantanoyl chloride, Hexahydro-2,5-methanopentalene-3a(1H)-carbonyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CZK 3,180.00

CZK 3,180.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
CZK 3,180.00

About This Item

Empirical Formula (Hill Notation):
C10H13ClO
CAS Number:
Molecular Weight:
184.66
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

CZK 3,180.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

solid

refractive index

n20/D 1.5159

density

1.165 g/mL at 25 °C

functional group

acyl chloride

SMILES string

ClC(=O)C12C[C@H]3C[C@H](C[C@H]1C3)C2

InChI

1S/C10H13ClO/c11-9(12)10-4-6-1-7(5-10)3-8(10)2-6/h6-8H,1-5H2/t6-,7+,8-,10-

InChI key

FEJBSNLESASJKD-WZXRXUJPSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

228.0 °F - closed cup

Flash Point(C)

108.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service