Skip to Content
Merck
All Photos(1)

Key Documents

675911

Sigma-Aldrich

N-Boc-indole-2-boronic acid

≥95%

Synonym(s):

1-(tert-butoxycarbonyl)indole-2-boronic acid, N-(tert-butoxycarbonyl)indole-2-boronic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CZK 1,510.00
5 G
CZK 7,190.00

CZK 1,510.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
CZK 1,510.00
5 G
CZK 7,190.00

About This Item

Empirical Formula (Hill Notation):
C13H16BNO4
CAS Number:
Molecular Weight:
261.08
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

CZK 1,510.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥95%

form

solid

mp

84-94 °C
85-90 °C (lit.)

storage temp.

−20°C

SMILES string

CC(C)(C)OC(=O)n1c(cc2ccccc12)B(O)O

InChI

1S/C13H16BNO4/c1-13(2,3)19-12(16)15-10-7-5-4-6-9(10)8-11(15)14(17)18/h4-8,17-18H,1-3H3

InChI key

SVIBPSNFXYUOFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

May contain varying amounts of anhydride

Application

Reactant involved in:
  • Suzuki-Miyaura cross-coupling reactions
  • Copper-catalyzed trifluoromethylation
  • Palladium-catalyzed benzylation
  • Homocoupling reactions
Used in a synthesis of hydroxyquinones via Suzuki-Miyaura coupling of phenylidonium ylides of hydroxyquinones.[1]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synlett, 2597-2597 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service