Skip to Content
Merck
All Photos(1)

Documents

670316

Sigma-Aldrich

Fmoc-N-(2-Boc-aminoethyl)-Gly-OH

≥97.0% (HPLC)

Synonym(s):

Fmoc-N-(2-Boc-aminoethyl)glycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H28N2O6
CAS Number:
Molecular Weight:
440.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97.0% (HPLC)

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Boc
Fmoc

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)NCCN(CC(O)=O)C(=O)OCC1c2ccccc2-c3ccccc13

InChI

1S/C24H28N2O6/c1-24(2,3)32-22(29)25-12-13-26(14-21(27)28)23(30)31-15-20-18-10-6-4-8-16(18)17-9-5-7-11-19(17)20/h4-11,20H,12-15H2,1-3H3,(H,25,29)(H,27,28)

InChI key

SMLJSDLXJRGOKW-UHFFFAOYSA-N

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service