Skip to Content
Merck
All Photos(1)

Key Documents

669407

Sigma-Aldrich

N-tert-Butyl-O-benzoylhydroxylamine hydrochloride

97%

Synonym(s):

Benzoic acid N-hydroxy-tert-butylamine ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)CO2NHC(CH3)3 HCl
Molecular Weight:
229.70
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

168 °C (dec.)

storage temp.

2-8°C

SMILES string

[Cl-].CC(C)(C)[NH2+]OC(=O)c1ccccc1

InChI

1S/C11H15NO2.ClH/c1-11(2,3)12-14-10(13)9-7-5-4-6-8-9;/h4-8,12H,1-3H3;1H

InChI key

UAYHNCIDRZTSPO-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service