Skip to Content
Merck
All Photos(1)

Documents

659738

Sigma-Aldrich

Poly[1,4-bis(hydroxyethyl)terephthalate-alt-ethyloxyphosphate]-co-1,4-bis(hydroxyethyl)terephthalate-co-terephthalate

Synonym(s):

P(BHET-EOP/BHET), 80/20

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(OCH2CH2OC(O)C6H4C(O)OCH2CH2OP(O)(OCH2CH3))x(OCH2CH2OC(O)C6H4C(O)OCH2CH2OC(O)C6H4C(O))y
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

solubility

THF: insoluble, but swells
chloroform: soluble
hexane: insoluble, but swells

General description

P(BHET-EOP/BHET) is a biodegradable polymer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service