Skip to Content
Merck
All Photos(1)

Key Documents

639192

Sigma-Aldrich

(3,3-Dimethyl-1-butynyl)boronic acid diisopropyl ester

97%

Synonym(s):

(2-tert-Butyl-1-ethynyl)diisopropoxyborane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H23BO2
CAS Number:
Molecular Weight:
210.12
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.4160 (lit.)

bp

193-194 °C (lit.)

density

0.891 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)OB(OC(C)C)C#CC(C)(C)C

InChI

1S/C12H23BO2/c1-10(2)14-13(15-11(3)4)9-8-12(5,6)7/h10-11H,1-7H3

InChI key

FLHZJMZNIOCRSX-UHFFFAOYSA-N

Application

Substrate that participates in a Dotz annulation process leading to a substituted arylboronate capable of further synthetic elaboration as for example, a Suzuki-Miyarura biaryl cross-coupling.[1]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

109.9 °F - closed cup

Flash Point(C)

43.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M W Davies et al.
The Journal of organic chemistry, 66(10), 3525-3532 (2001-05-12)
This paper describes the synthesis and reactivity of a novel class of quinone boronic esters. These compounds are prepared utilizing a highly regioselective Dötz annulation of Fischer carbene complexes with alkynylboronates. All substrates studied to date provided a single regioisomeric

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service