Recommended Products
Assay
97%
refractive index
n20/D 1.4990 (lit.)
bp
183-184 °C (lit.)
density
1.015 g/mL at 25 °C (lit.)
SMILES string
CNCc1cccc(F)c1
InChI
1S/C8H10FN/c1-10-6-7-3-2-4-8(9)5-7/h2-5,10H,6H2,1H3
InChI key
ZXWCKKSSCIFVBT-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
152.1 °F
Flash Point(C)
66.7 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Dalton transactions (Cambridge, England : 2003), 49(21), 7252-7257 (2020-05-20)
Organic-inorganic hybrid materials with perovskite structure have recently attracted tremendous interest due to their structural tunability and rich functional properties, such as phase transition and photoelectric properties. Within this context, two discrete tin-based organic-inorganic hybrid compounds [(FMBA)2SnCl6] (1) and [(FMBA)2SnBr6]
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service