Skip to Content
Merck
All Photos(1)

Key Documents

61897

Sigma-Aldrich

Ethyl piperazinoacetate

≥95.0% (GC)

Synonym(s):

1-(Ethoxycarbonylmethyl)piperazine, N-(Carboethoxymethyl)-piperazine, Ethyl 1-piperazineacetate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
CZK 3,210.00

CZK 3,210.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
CZK 3,210.00

About This Item

Empirical Formula (Hill Notation):
C8H16N2O2
CAS Number:
Molecular Weight:
172.22
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

CZK 3,210.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥95.0% (GC)

reaction suitability

reaction type: solution phase peptide synthesis

refractive index

n20/D 1.472

density

1.048 g/mL at 20 °C (lit.)

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

CCOC(=O)CN1CCNCC1

InChI

1S/C8H16N2O2/c1-2-12-8(11)7-10-5-3-9-4-6-10/h9H,2-7H2,1H3

InChI key

MTFCXMJOGMHYAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Warning

Irritant

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service