Skip to Content
Merck
All Photos(1)

Documents

610011

Sigma-Aldrich

Boron-11B trifluoride

≥95 atom % 11B

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
11BF3
CAS Number:
Molecular Weight:
68.00
EC Number:
MDL number:

isotopic purity

≥95 atom % 11B

form

gas

mol wt

67.96 by atom % calculation

bp

-100 °C (lit.)

mp

-127 °C (lit.)

mass shift

depleted

InChI

1S/BF3/c2-1(3)4

InChI key

WTEOIRVLGSZEPR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service