Skip to Content
Merck
All Photos(1)

Documents

604984

Sigma-Aldrich

DL-Glutamic acid-13C5

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO213C(13CH2)213CH(NH2)13CO2H
Molecular Weight:
152.09
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

99 atom % 13C

mp

185 °C (dec.) (lit.)

mass shift

M+5

SMILES string

N[13CH]([13CH2][13CH2][13C](O)=O)[13C](O)=O

InChI

1S/C5H9NO4/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/i1+1,2+1,3+1,4+1,5+1

InChI key

WHUUTDBJXJRKMK-CVMUNTFWSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service