Skip to Content
Merck

596183

3-Bromo-2-methoxy-5-methylphenylboronic acid

Synonym(s):

3-Bromo-5-methyl-2-methoxyphenylboronic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H10BBrO3
CAS Number:
Molecular Weight:
244.88
UNSPSC Code:
12352103
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


mp

102-106 °C (lit.)

SMILES string

COc1c(Br)cc(C)cc1B(O)O

InChI

1S/C8H10BBrO3/c1-5-3-6(9(11)12)8(13-2)7(10)4-5/h3-4,11-12H,1-2H3

InChI key

WHQCYDMSPACJBE-UHFFFAOYSA-N

Other Notes

Contains varying amounts of anhydride


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
596183-1G04061825959184