Skip to Content
Merck
All Photos(1)

Key Documents

595861

Sigma-Aldrich

PAMAM dendrimer, 1,4-diaminobutane core, generation 1.0 solution

20 wt. % in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH2(CH2)4NH2]:(G=1);dendri PAMAM(NH2)8
Molecular Weight:
1457.90
MDL number:
UNSPSC Code:
12162002

description

amino surface groups

form

liquid

mol wt

generation 1

feature

core type 1,4-diaminobutane core (4-carbon core)

concentration

20 wt. % in methanol

no. Surface Groups

8

refractive index

n20/D 1.370

bp

64.7 °C

density

0.791 g/mL at 25 °C

storage temp.

−20°C

Features and Benefits

Contains 8 surface amino groups.

Packaging

unit weight includes solvent

Legal Information

Mfg. by Andrews ChemServices. Licensed for research use only.
Mfg. by Dendritic Nanotechnologies Inc. Licensed for research use only.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service