Skip to Content
Merck
All Photos(1)

Key Documents

588059

Sigma-Aldrich

Chloroethane-1,1-d2

98 atom % D

Synonym(s):

Ethyl-1,1-d2 chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CD2Cl
CAS Number:
Molecular Weight:
66.53
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.12

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % D

bp

12.3 °C (lit.)

mass shift

M+2

SMILES string

[2H]C([2H])(C)Cl

InChI

1S/C2H5Cl/c1-2-3/h2H2,1H3/i2D2

InChI key

HRYZWHHZPQKTII-CBTSVUPCSA-N

Related Categories

Packaging

Standard 2.5 g quantity is packaged in a 450 mL carbon steel lecture bottle with brass CGA 110/180 valve. Nominal gas pressure at 21ºC is 5 psig for 2.5 g. This pressure is slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Stainless steel control valves Z146951 or Z187224
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Flam. Gas 1A - Press. Gas Liquefied gas - Repr. 1B

Storage Class Code

2A - Gases

WGK

WGK 2

Flash Point(F)

-58.0 °F - closed cup

Flash Point(C)

-50.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service