Skip to Content
Merck
All Photos(1)

Documents

586641

Sigma-Aldrich

Acrylonitrile-13C3

≥99 atom % 13C, ≥99% (CP), contains hydroquinone as stabilizer

Synonym(s):

2-Propenenitrile-1,2,3-13C3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H213C=13CH13CN
CAS Number:
Molecular Weight:
56.04
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

≥99 atom % 13C

Quality Level

Assay

≥99% (CP)

contains

hydroquinone as stabilizer

bp

77 °C (lit.)

mp

-83 °C (lit.)

density

0.850 g/mL at 25 °C

mass shift

M+3

storage temp.

2-8°C

SMILES string

[13CH2]=[13CH][13C]#N

InChI

1S/C3H3N/c1-2-3-4/h2H,1H2/i1+1,2+1,3+1

InChI key

NLHHRLWOUZZQLW-VMIGTVKRSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

23.0 °F - closed cup

Flash Point(C)

-4.99 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shun-Suke Moriya et al.
Analytical biochemistry, 607, 113831-113831 (2020-08-03)
We developed a new procedure for the comprehensive analysis of metabolites and enzymes involved in polyamine metabolism pathways. The procedure utilizes stable isotope-labeled polyamines and directly and precisely determines labeled products from enzymatic reactions by ESI-Q-TOF-MS. The activity of different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service