Skip to Content
Merck
All Photos(1)

Key Documents

572446

Sigma-Aldrich

3-Methyl-4-nitrobenzyl bromide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)CH2Br
CAS Number:
Molecular Weight:
230.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

97%

mp

46-48 °C (lit.)

SMILES string

Cc1cc(CBr)ccc1[N+]([O-])=O

InChI

1S/C8H8BrNO2/c1-6-4-7(5-9)2-3-8(6)10(11)12/h2-4H,5H2,1H3

InChI key

WOUWGCLQOJPWHS-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service