567655
5-Methoxy-3-indolylacetonitrile
95%
Synonym(s):
3-(Cyanomethyl)-5-methoxyindole
About This Item
Assay
95%
mp
65-69 °C (lit.)
SMILES string
COc1ccc2[nH]cc(CC#N)c2c1
InChI
1S/C11H10N2O/c1-14-9-2-3-11-10(6-9)8(4-5-12)7-13-11/h2-3,6-7,13H,4H2,1H3
InChI key
ZBQCXEREMRGOCO-UHFFFAOYSA-N
Application
Reactant for preparation of carboline analogs as potent MAPKAP-K2 inhibitors
Reactant for preparation of indole-N-acetic acid derivatives as aldose reductase inhibitors for diabetic complications treatment
Reactant for preparation of debromoflustramine B and labeled Me(3a)-13C-physostigmine
Reactant for preparation of physostigmine and physovenine
Reactant for preparation of phenethyl substituted indole derivatives as melatoninergic agonists and antagonists
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service