Recommended Products
Assay
97%
mp
53-57 °C (lit.)
SMILES string
Cc1cc(ccc1Br)C#N
InChI
1S/C8H6BrN/c1-6-4-7(5-10)2-3-8(6)9/h2-4H,1H3
InChI key
SKXUZFJOLNNWIG-UHFFFAOYSA-N
Application
4-Bromo-3-methylbenzonitrile may be used to synthesize 2,4,6-tris(4-bromo-3-methyl-phenylene)-1,3,5-triazine and 4-fluoro-3-methylbenzonitrile.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
"The Evolution of Pd0/PdII-Catalyzed Aromatic Fluorination"
Accounts of Chemical Research, 49(10), 2146-2157 (2016)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service