Skip to Content
Merck
All Photos(1)

Documents

557994

Sigma-Aldrich

2-Fluorothioanisole

97%

Synonym(s):

2-Fluorophenyl methyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4SCH3
CAS Number:
Molecular Weight:
142.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.5590 (lit.)

bp

197-199 °C (lit.)

density

1.174 g/mL at 25 °C (lit.)

SMILES string

CSc1ccccc1F

InChI

1S/C7H7FS/c1-9-7-5-3-2-4-6(7)8/h2-5H,1H3

InChI key

AYTSELVZWGHUEE-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

195.1 °F - closed cup

Flash Point(C)

90.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jean Sun Lim et al.
Chemical science, 10(8), 2404-2412 (2019-03-19)
The surface crossing of bound and unbound electronic states in multidimensional space often gives rise to resonances in the continuum. This situation happens in the πσ*-mediated photodissociation reaction of 2-fluorothioanisole; optically-bright bound S1 (ππ*) vibrational states of 2-fluorothioanisole are strongly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service