Skip to Content
Merck
All Photos(1)

Key Documents

55761

Sigma-Aldrich

(Hydroxymethyl)polystyrene

200-500 mesh, extent of labeling: ~1.1 mmol/g loading

Synonym(s):

Benzyl alcohol polymer-bound, Polystyrene crosslinked with divinylbenzene, hydroxymethylated

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reaction type: Boc solid-phase peptide synthesis
reactivity: carboxyl reactive

extent of labeling

~1.1 mmol/g loading

matrix

crosslinked with 1% DVB

particle size

200-500 mesh

functional group

alcohol

InChI

1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2

InChI key

WVDDGKGOMKODPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service