Skip to Content
Merck
All Photos(1)

Key Documents

55609

Sigma-Aldrich

17β-Hydroxy-17-methylandrosta-1,4-dien-3-one

≥99.0% (HPLC)

Synonym(s):

1-Dehydro-17α-methyltestosterone, Dianabol, Methandienone, Methandrostenolone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28O2
CAS Number:
Molecular Weight:
300.44
Beilstein:
2056255
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:

Assay

≥99.0% (HPLC)

form

solid

optical activity

[α]20/D −0.5±0.5°, c = 1.1% in chloroform

drug control

USDEA Schedule IIIN; regulated under CDSA - not available from Sigma-Aldrich Canada

mp

164-166 °C

storage temp.

2-8°C

SMILES string

C[C@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C20H28O2/c1-18-9-6-14(21)12-13(18)4-5-15-16(18)7-10-19(2)17(15)8-11-20(19,3)22/h6,9,12,15-17,22H,4-5,7-8,10-11H2,1-3H3/t15-,16+,17+,18+,19+,20+/m1/s1

InChI key

XWALNWXLMVGSFR-HLXURNFRSA-N

Gene Information

human ... AR(367)

Looking for similar products? Visit Product Comparison Guide

Application

17β-Hydroxy-17-methylandrosta-1,4-dien-3-one can be used as a model steroid in the screening of androgenic anabolic steroids using MALDI-TOF technique.

Other Notes

Sales restrictions may apply

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparative study of matrices for their use in the rapid screening of anabolic steroids by matrix-assisted laser desorption/ionisation time-of-flight mass spectrometry
Galesio M, et al.
Rapid Communications in Mass Spectrometry, 23(12), 1783-1791 (2009)
Stephanie Mosler et al.
Archives of toxicology, 86(1), 109-119 (2011-08-06)
There is increasing evidence that the biological activity of myostatin (MSTN), a negative regulator of muscle growth, is affected by training but also anabolic steroids. In this study, we analyzed the effects of the frequently abused anabolic steroid methandienone (Md)
Linda L Ahonen et al.
Rapid communications in mass spectrometry : RCM, 24(7), 958-964 (2010-03-09)
We examined the feasibility of capillary liquid chromatography/microchip atmospheric pressure photoionization tandem mass spectrometry (capLC/microAPPI-MS/MS) for the analysis of anabolic steroids in human urine. The urine samples were pretreated by enzymatic hydrolysis (with beta-glucuronidase from Helix pomatia), and the compounds
Oscar J Pozo et al.
Drug testing and analysis, 1(11-12), 554-567 (2010-04-01)
Anabolic steroids are among the most frequently detected compounds in doping analysis. They are extensively metabolized and therefore an in-depth knowledge about steroid metabolism is needed. In this study, a liquid chromatography tandem mass spectometry (LC-MS/MS) method based on a
Yun Wang et al.
International journal of biological macromolecules, 49(3), 428-432 (2011-06-15)
The development of a chitosan-supported immunoaffinity chromatography (IAC) column and its application to the selective extraction of methandrostenolone (MA) from food and feed samples were described in this paper. Using hybridoma technique, a monoclonal antibody (mAb) against MA was produced.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service