Skip to Content
Merck
All Photos(1)

Key Documents

555770

Sigma-Aldrich

α,α,p-Tribromoacetophenone

97%

Synonym(s):

2,2,4′-Tribromoacetophenone, 2,2-Dibromo-1-(4-bromophenyl)ethanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br2CHCOC6H4Br
CAS Number:
Molecular Weight:
356.84
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
To inquire about this product 555770, please contact your local Merck office or dealer. Contact Technical Service

Assay

97%

form

solid

mp

91-94 °C (lit.)

SMILES string

BrC(Br)C(=O)c1ccc(Br)cc1

InChI

1S/C8H5Br3O/c9-6-3-1-5(2-4-6)7(12)8(10)11/h1-4,8H

InChI key

KFTUNOVZJWIKFX-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service