Skip to Content
Merck
All Photos(1)

Key Documents

555142

Sigma-Aldrich

1-Diphenylphosphino-1′-(di-tert-butylphosphino)ferrocene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CZK 4,480.00

CZK 4,480.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
CZK 4,480.00

About This Item

Empirical Formula (Hill Notation):
C30H36FeP2
CAS Number:
Molecular Weight:
514.40
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

CZK 4,480.00


Please contact Customer Service for Availability

Request a Bulk Order

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
reaction type: Coupling Reactions

mp

75-79 °C (lit.)

functional group

phosphine

SMILES string

[Fe].CC(C)(C)P([C]1[CH][CH][CH][CH]1)C(C)(C)C.[CH]2[CH][CH][C]([CH]2)P(c3ccccc3)c4ccccc4

InChI

1S/C17H14P.C13H22P.Fe/c1-3-9-15(10-4-1)18(17-13-7-8-14-17)16-11-5-2-6-12-16;1-12(2,3)14(13(4,5)6)11-9-7-8-10-11;/h1-14H;7-10H,1-6H3;

InChI key

GQIMIFFIUZZVMJ-UHFFFAOYSA-N

Application

  • Catalyst in microwave-assisted preparation of aryl-alkyl ethers via palladium-catalyzed Hiyama coupling of alkoxysilanes with aryl bromides or aryl chlorides
  • Ligand for catalytic isomerization of methylbutenenitrile via nickel systems
  • Ligand for ruthenium alkylidene complexes for catalysis of metathesis reactions and catalytic norbornene polymerization

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marvin Parasram et al.
Journal of the American Chemical Society, 136(52), 17926-17929 (2014-12-17)
A palladium (Pd)-catalyzed endo-selective Heck reaction of iodomethylsilyl ethers of phenols and aliphatic alkenols has been developed. Mechanistic studies reveal that this silyl methyl Heck reaction operates via a hybrid Pd-radical process and that the silicon atom is crucial for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service